Learn More
BMS-193885, MedChemExpress
Código de producto. 16434968
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
Unit Size:
10 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16434968 10 mg 10 miligramos
16407707 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16434968 Supplier Medchem Express Supplier No. HY120619
This item is not returnable. View return policy
MedChemExpress BMS-193885 is a potent, selective, competitive, and brain penetrant neuropeptide Y1 receptor antagonist with a Ki of 3.3 nM, and has an IC50 of 5.9 nM for hY1, which displays > 100, > 160, > 160 and > 160-fold selectivity over α1, hY2, hY4 and hY5 receptors, respectively .
BMS-193885 is a potent, selective, competitive, and brain penetrant neuropeptide Y1 receptor antagonist with a Ki of 3.3 nM, and has an IC50 of 5.9 nM for hY1, which displays > 100, > 160, > 160 and > 160-fold selectivity over α1, hY2, hY4 and hY5 receptors, respectively .
Specifications
Nombre del producto químico o material BMS-193885
CAS 186185-03-5
Color Naranja
Notas de grado de pureza Research
Fórmula molecular C33H42N4O6
Cantidad 10 mg
Información de solubilidad DMSO : 100 mg/mL (169.29 mM; Need ultrasonic)
SMILES O=C(C1=C(C)NC(C)=C(C(OC)=O)C1C2=CC=CC(NC(NCCCN3CCC(C4=CC=CC(OC)=C4)CC3)=O)=C2)OC
Peso molecular (g/mol) 590.71
Formula Weight (peso de la fórmula) 590.71
Porcentaje de pureza 97.73%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Neuroscience-Neuromodulation