Learn More
Tocris Bioscience™ BMH 21
Descripción
BMH 21 is a p53 pathway activator; induces expression of p53 target genes. Also RNA polymerase 1 inhibitor; promotes degradation of RPA194. Reduces viability of multiple tumor cell lines and attenuates tumor growth of A375 melanoma cell xenografts in mice.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-[2-(Dimethylamino)ethyl]-12-oxo-12 H-benzo[g]pyrido[2,1-b]quinazoline-4-carboxamide |
| CAS | 896705-16-1 |
| Cantidad | 10 mg |
| Objetivo | RNA/DNA Polymerase Inhibitors |
| Fórmula molecular | C21H20N4O2 |
| Formula Weight (peso de la fórmula) | 360.41 |
| Pureza | 98% |