Learn More
Tocris Bioscience™ BIX NHE1 inhibitor
Descripción
BIX NHE1 inhibitor is a potent and selective NHE1 inhibitor (IC50 = 31 nM). Selective for NHE1 over NHE2 and NHE3. Prevents ischemic damage in an ischemia reperfusion injury isolated rat heart model ex vivo. Prevents phenylephrine -induced cardiomyocyte hypertrophy in vitro, and attenuates cardiac hypertrophy and left ventricular dysfunction postinfarction in rats. Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-(1-Acetyl-4-piperidinyl)-N-(aminoiminomethyl)-3-(trifluoromethyl)benzamide hydrochloride |
| CAS | 1422252-46-7 |
| Cantidad | 50 mg |
| Objetivo | Na+/Ca2+ Exchanger Inhibitors |
| Fórmula molecular | C16H19F3N4O2.HCl |
| Formula Weight (peso de la fórmula) | 392.8 |
| Pureza | >98% |