Learn More
Agilent BioTek Filters for ELx800 Absorbance Microplate Reader
Accessory for use with ELx800 Absorbance Microplate Reader.
Brand: Agilent BioTek 7334650
This item is not returnable.
View return policy
Description
Accessory for use with ELx800 Absorbance Microplate Reader.
Specifications
Filter | |
ELx800 Absorbance Microplate Readers | |
650 nm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Agilent BioTek Filters for ELx800 Absorbance Microplate Reader > 650nm
Spot an opportunity for improvement?Share a Content Correction