Descripción
- 4 in. deep corrosion-resistant polyurethane bin for use with the Flexi-Bin Cart
- Exterior dimensions: 22.25 x 17 x 4 in.
- Interior dimensions: 20.25 x 15.25 x 3.8 in.
- Includes three long (20.25 in.) and four short (15.25 in.) polyurethane dividers for partitioning the bin to securely hold items in upright positions
- 10 in. deep bin is also available
Especificaciones
Especificaciones
| Tipo de producto | Replacement Bin |
| Material | Polyethylene |