Learn More
Bif-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16874584 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16874584 20 μL 20 microlitros
16864584 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16874584 Proveedor Proteintech N.º de proveedor 154221AP20UL
Rabbit Polyclonal Antibody
The Bcl-2 family consists of anti-apoptotic (or cell survival) genes, such as Bcl-2, Bcl-XL and pro-apoptotic (or cell death) genes including Bax and BAD. In addition to its anti-apoptotic effect, Bcl-2 also inhibits progression through the cell cycle. Functional interactions between family members as well as binding to other cellular proteins modulate their activities. Bcl-2 family members are important in normal tissue development, homeostasis, and disease states. Bcl-2 prevents cell death by forming heterodimers with Bax and Bad (1). Bax promotes cell death by forming homodimers, which can be blocked by forming heterodimers with Bcl-2 or BCL-XL. Recently, using yeast two-hybrid system Cuddeback et al (product citation 2) have identified a cDNA encoding a 365 amino acid protein, which interacts physically with Bax protein and influences cell life and death. Bif-1 contains a Src homology 3 (SH3) domain in the C-terminus. It does not contain any of the conserved Bcl-2 homology (BH) domains of the Bcl-2 family proteins. Human Bif-1 protein shares 96% and 42% identity at the amino acids level with mouse and C. elegans, respectively. Overexpression of Bif-1 in transfected cell lines promotes Bax conformational change, caspase activation, and apoptotic cell death following growth factor withdrawal. Bif-1 RNA is abundantly expressed in heart, skeletal muscle, kidney, and placenta.
Especificaciones
Antígeno Bif-1
Aplicaciones Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.29 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica SH3GLB1
N.º de referencia del gen Q6AYE2, Q9JK48, Q9Y371
Alias de gen Bax interacting factor 1, Bif 1, Bif1, Endophilin B1, SH3GLB1
Símbolos de los genes SH3GLB1
Especie del huésped Rabbit
Inmunógeno Bif-1 Fusion Protein Ag7710
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 292156, 51100, 54673
Especies diana Mouse, Rat, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG