Learn More
BI-9627, MedChemExpress
Descripción
BI-9627 displays >30-fold selectivity against NHE2 and with no measurable inhibitory activity against the NHE3 isoform. BI-9627 shows low DDI (drug-drug interaction) potential, excellent pharmacokinetics in rat and dog, and remarkably potent activity in the isolated heart model of ischemia-reperfusion injury.
Especificaciones
Especificaciones
| Nombre del producto químico o material | BI-9627 |
| CAS | 1204329-34-9 |
| Color | Yellow |
| Notas de grado de pureza | Research |
| Fórmula molecular | C16H19F3N4O2 |
| Cantidad | 5 mg |
| Información de solubilidad | DMSO : 100 mg/mL (280.63 mM; Need ultrasonic) |
| SMILES | NC(NC(C1=CC=C(C2CCN(C(C)=O)CC2)C(C(F)(F)F)=C1)=O)=N |
| Peso molecular (g/mol) | 356.34 |
| Formula Weight (peso de la fórmula) | 356.34 |
| Mostrar más |