Learn More
Tocris Bioscience™ BI 9564
Potent and selective BRD9 and BRD7 inhibitor; orally active
Marca: Tocris Bioscience™ 5590/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
BI 9564 is a potent and selective BRD9 and BRD7 inhibitor (Kd values are 14 and 239 nM, respectively). Exhibits selectivity for BRD9/7 over 48 other bromodomains, 324 kinases and 55 GPCRs. Inhibits proliferation of AML cell lines in vitro and reduces AML tumor xenograft growth in vivo. Cell permeable and orally active.Especificaciones
| 4-[4-[(Dimethylamino)methyl]-2,5-dimethoxyphenyl]-2-methyl-1,2-dihydro-2,7-naphthyridin-1-one | |
| 1883429-22-8 | |
| 10 mg |