Learn More
BI-9564, MedChemExpress
Código de producto. 16499526
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
2 mg
5 mg
10 mg
50 mg
100 mg
200 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
2 miligramos
200 miligramos
5 miligramos
50 miligramos
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
16499526 50 mg 50 miligramos
16494276 100 mg 100 miligramos
16490006 2 mg 2 miligramos
16482958 10 mg 10 miligramos
16480906 200 mg 200 miligramos
16405707 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Código de producto. 16499526 Proveedor Medchem Express N.º de proveedor HY100352
MedChemExpress BI-9564 is a potent, selective and cell-permeable BRD9/BRD7 bromodomains inhibitor, with IC50s of 75 nM and 3.4 μM and Kds of 14 nM and 239 nM, respectively. BI-9564 has an IC50 of > 100 μM for BET family.
BI-9564 is a potent, selective and cell-permeable BRD9/BRD7 bromodomains inhibitor, with IC50s of 75 nM and 3.4 μM and Kds of 14 nM and 239 nM, respectively. BI-9564 has an IC50 of > 100 μM for BET family.
Especificaciones
Nombre del producto químico o material BI-9564
CAS 1883429-22-8
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C20H23N3O3
Cantidad 50 mg
Información de solubilidad DMSO : 8.33 mg/mL (23.57 mM; Need ultrasonic)
SMILES CN(C)CC1=CC(OC)=C(C(C2=C3C=NC=C2)=CN(C)C3=O)C=C1OC
Peso molecular (g/mol) 353.41
Formula Weight (peso de la fórmula) 353.41
Porcentaje de pureza 99.86%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease