Learn More
Betamethasone dipropionate, MedChemExpress
MedChemExpress Betamethasone dipropionate is a glucocorticoid steroid with anti-inflammatory and immunosuppressive abilities.
Marca: Medchem Express HY-13571
Descripción
Betamethasone dipropionate is a glucocorticoid steroid with anti-inflammatory and immunosuppressive abilities.Especificaciones
Betamethasone dipropionate | |
Blanco | |
C28H37FO7 | |
Betamethasone 17,21-dipropionate | |
C[C@@]12[C@](C[C@H](C)[C@]2(OC(CC)=O)C(COC(CC)=O)=O)([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@@]3(F)[C@@H](O)C1)=O | |
504.59 | |
Research | |
COVID-19-immunoregulation |
5593-20-4 | |
Research | |
250 mg | |
DMSO : 110 mg/mL (218.00 mM; Need ultrasonic) | |
504.59 | |
98.31% | |
Solid |
Sugerencias de productos
Customers who viewed this item also viewed.