Learn More
beta-Cyfluthrin, TRC
Código de producto. 30134886
Change view
Click to view available options
Cantidad:
1 g
5 g
25 g
Tamaño de la unidad:
1 g
25 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30134886 25 g 25 g
30134488 5 g 5 g
17917539 1 g 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30134886 Proveedor Toronto Research Chemicals N.º de proveedor TRCC98951025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
CAS 68359-37-5
Fórmula molecular C22 H18 Cl2 F N O3
Peso molecular (g/mol) 434.29
Sinónimo BAY-FCR 1272, BAY-Vl 1704, Baythroid, Cyfloxylate, Cyfluthrin, Eulan SP, FCR 1272, Optem PT 600, Preventol HS 12, Renounce, Solfac, Solfac 050EW, Tempo 2, Tempo 20WP, Tombstone, Tombstone Helios, α-Cyano-3-phenoxy-4-fluorobenzyl 2,2-dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarboxylate, α-Cyano-4-fluoro-3-phenoxybenzyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate
Nombre IUPAC [cyano-(4-fluoro-3-phenoxyphenyl)methyl] 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
SMILES CC1(C)C(C=C(Cl)Cl)C1C(=O)OC(C#N)c2ccc(F)c(Oc3ccccc3)c2
Especificaciones
Nombre del producto químico o material Cyfluthrin
Notas de grado de pureza HPLC
Cantidad 25 g
Fórmula InChI InChI=1S/C22H18Cl2FNO3/c1-22(2)15(11-19(23)24)20(22)21(27)29-18(12-26)13-8-9-16(25)17(10-13)28-14-6-4-3-5-7-14/h3-11,15,18,20H,1-2H3
Formula Weight (peso de la fórmula) 433.0648
Porcentaje de pureza >95%