Learn More
Bromuro de beta-ciclogeraniltrifenilfosfonio-d5, TRC
Product Code. 30352561
Change view
Click to view available options
Cantidad:
5 mg
Unit Size:
5 miligramos
Product Code. Cantidad unitSize
30352561 5 mg 5 miligramos
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30352561 Supplier Toronto Research Chemicals Supplier No. TRCC9878525MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material beta-Cyclogeranyltriphenylphosphonium Bromide-d5
CAS 78995-97-8
Fórmula molecular C28H27D5BrP
Cantidad 5 mg
Sinónimo Triphenyl[(2,6,6-trimethyl-1-cyclohexen-1-yl)methyl]phosphonium Bromide-d5
Fórmula InChI InChI=1S/C28H32P.BrH/c1-23-14-13-21-28(2,3)27(23)22-29(24-15-7-4-8-16-24,25-17-9-5-10-18-25)26-19-11-6-12-20-26;/h4-12,15-20H,13-14,21-22H2,1-3H3;1H/q+1;/p-1/i1D3,14D2;
SMILES [Br-].[2H]C([2H])([2H])C1=C(C[P+](c2ccccc2)(c3ccccc3)c4ccccc4)C(C)(C)CCC1([2H])[2H]
Nombre IUPAC Bromuro de [3,3-dideuterio-6,6-dimetil-2-(trideuteriometil)ciclohexen-1-il]metil-trifenilfosfanio
Peso molecular (g/mol) 484.46
Formula Weight (peso de la fórmula) 483.1739