Learn More
beta Actin Mouse anti-Hamster, Human, Mouse, Non-human primate, Rat, Zebrafish, Clone: 2D4H5, Proteintech
Código de producto. 16817505 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
Tamaño de la unidad:
150 microlitros
16817505 150 μL 150 microlitros
1 options
Código de producto. 16817505 Proveedor Proteintech N.º de proveedor 660091IG150UL
Mouse Monoclonal Antibody
This antibody can recognize all the actins. The isotype of this antibody is IgG2b.
The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
Especificaciones
Antígeno beta Actin
Aplicaciones Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Clasificación Monoclonal
Clon 2D4H5
Concentración 0.43 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica ACTA1
N.º de referencia del gen P48975, P60709, P60710, P60711, P62736, P62737, P62738, P63259, P63260, P63261, P63267, P63268, P63269, P68032, P68033, P68035, P68133, P68134, P68136
Alias de gen ACTB, Actin, actin, beta, Actin, cytoplasmic 1, B actin, Beta actin, F actin, PS1TP5BP1
Símbolos de los genes ACTA1, acta1b, acta2, ACTB, actb1, ACTC1, actc1b, Actg1, ACTG2
Especie del huésped Mouse
Método de purificación Protein A
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100689477, 100758575, 100764022, 100766388, 100768744, 100768972, 11459, 11461, 11464, 11465, 11468, 11475, 25365, 287876, 29275, 29437, 322509, 407658, 57934, 58, 58114, 59, 60, 70, 71, 72, 81633, 81822
Especies diana Hamster, Human, Mouse, Non-human Primate, Rat, Zebrafish
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b