Learn More
BD Phosflow™ Human Monocyte/NK Cell Activation Kit
Descripción
Includes
Antibody cocktail, fluorochrome-labled antibodies to six key phosphoproteins, optimized protocols and buffers
Complete system for the determination of the activation status of six different key phosphoproteins in B cells, monocytes, NK cells, and T cells using whole blood samples.
Evaluate:
- Phosphorylation status information on the intracellular MAPK and JAK/STAT signaling pathway
- Responses transduced by treated human monocytes and NK cells
- Signaling events in T and B-cell populations
- Lyophilized positive and negative control cells to increase confidence and standardization
Flow cytometry
Especificaciones
Especificaciones
| Especie del huésped | Human |
| Cantidad | 50 Tests |
| Tipo de producto | Human Monocyte/NK Cell Activation Kit |