Learn More
Tocris Bioscience™ BAY 593
Descripción
BAY 593 is a geranylgeranyltransferase-I (GGTase-I) inhibitor. Blocks activation of Rho-GTPases to inactivate YAP1/TAZ signaling. Dose-dependently inhibits growth of tumors in xenograft mouse models (IC50 values are 38.4 nM and 564 nM in MT-1080 and MDA-MB-231 cells respectively) and PXF 541 xenografts. Orally bioavailable and suitable for use in vivo.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (2R)-3,3,3-Trifluoro-2-methoxy-1-[(7R)-7-(2-methoxyphenyl)-3,9-diazaspiro[5.5]undec-3-yl]-2-phenyl-1-propanone hydrochloride |
| CAS | 2413020-57-0 |
| Cantidad | 5 mg |
| Objetivo | Other Transferase Inhibitors |
| Fórmula molecular | C26H31F3N2O3.HCl |
| Pureza | 98% |