Learn More
Tocris Bioscience™ Bay 36-7620
Descripción
Selective mGlu1 receptor non-competitive antagonist (IC50 = 0.16 μM) with inverse agonist activity. Impairs classical conditioning and associated synaptic plasticity in hippocampal neurons. Exhibits neuroprotective and anticonvulsive effects in vivo following systemic administration.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Bay 36-7620 |
| CAS | 232605-26-4 |
| Cantidad | 10 mg |
| Fórmula molecular | C19H18O2 |
| Formula Weight (peso de la fórmula) | 278.35 |
| Pureza | >98% |
| Peso molecular | 278.351 |