Learn More
Balicatib, MedChemExpress
Código de producto. 16488897
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16488897 5 mg 5 miligramos
16436636 100 mg 100 miligramos
16436318 10 mg 10 miligramos
16402437 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16488897 Proveedor Medchem Express N.º de proveedor HY15100
MedChemExpress Balicatib (AAE581) is a potent, orally active and selective cathepsin K inhibitor with IC50 values of 22, 61, 48, 2900 nM for cathepsin K, cathepsin B, cathepsin L, cathepsin S, respectively. Balicatib inhibits bone turnover, decreases bone formation rates. Balicatib has the potential for the research of osteoporosis.
Balicatib (AAE581) is a potent, orally active and selective cathepsin K inhibitor with IC50 values of 22, 61, 48, 2900 nM for cathepsin K, cathepsin B, cathepsin L, cathepsin S, respectively. Balicatib inhibits bone turnover, decreases bone formation rates. Balicatib has the potential for the research of osteoporosis.
Especificaciones
Nombre del producto químico o material Balicatib
Forma física Solid
CAS 354813-19-7
Color Blanco
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C23H33N5O2
Para utilizar con (aplicación) Neuroscience-Neuromodulation
Sinónimo AAE581
Información de solubilidad DMSO : 75 mg/mL (182.24 mM; Need ultrasonic)
SMILES CCCN1CCN(C2=CC=C(C(NC3(C(NCC#N)=O)CCCCC3)=O)C=C2)CC1
Peso molecular (g/mol) 411.54
Formula Weight (peso de la fórmula) 411.54
Porcentaje de pureza 99.07%