Learn More
Éster metílico de azilsartán-d5, TRC
Product Code. 30332259
Change view
Click to view available options
Cantidad:
10 mg
Unit Size:
10 miligramos
Product Code. Cantidad unitSize
30332259 10 mg 10 miligramos
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30332259 Supplier Toronto Research Chemicals Supplier No. TRCA92690710MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Azilsartan-d5 Methyl Ester
CAS 1794766-42-9
Fórmula molecular C26H17D5N4O5
Cantidad 10 mg
Sinónimo 1-[[2'-(2,5-Dihydro-5-oxo-1,2,4-oxadiazol-3-yl)[1,1'-biphenyl]-4-yl]methyl]-2-ethoxy-1H-benzimidazole-7-carboxylic-d5 Acid Methyl Ester
Fórmula InChI InChI=1S/C26H22N4O5/c1-3-34-25-27-21-10-6-9-20(24(31)33-2)22(21)30(25)15-16-11-13-17(14-12-16)18-7-4-5-8-19(18)23-28-26(32)35-29-23/h4-14H,3,15H2,1-2H3,(H,28,29,32)/i1D3,3D2
SMILES [2H]C([2H])([2H])C([2H])([2H])Oc1nc2cccc(C(=O)OC)c2n1Cc3ccc(cc3)c4ccccc4C5=NC(=O)ON5
Nombre IUPAC 3-[[4-[2-(5-oxo-2H-1,2,4-oxadiazol-3-il)fenil]fenil]metil]-2-(1,1,2,2,2-pentadeuterioetoxi)benzimidazol-4-carboxilato de metilo
Peso molecular (g/mol) 475.51
Formula Weight (peso de la fórmula) 475.1904