Learn More
Conjugado de azido-PEG3-sulforodamina101, Thermo Scientific Chemicals
Código de producto. 15456919
Cambiar vista
Click to view available options
Cantidad:
5 mg
25 mg
Tamaño de la unidad:
25 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
15456919 5 mg 5 miligramos
15466919 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 15456919 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor J65476.#0
Solicitar formato a granel o personalizado
Una sulforodamina 101 soluble en agua con funcionalidad de grupo azida
A water soluble sulforhodamine 101 with azido group functionality
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 1353153-76-0
Fórmula molecular C39H46N6O9S2
Número MDL MFCD22380752
Sinónimo Azide-Fluor 585
Especificaciones
Nombre del producto químico o material Azido-PEG3-sulforhodamine 101 conjugate
Color Rojo
Cantidad 5 mg
Formula Weight (peso de la fórmula) 806.95
Forma física Sólido