Learn More
AZD9567, MedChemExpress
Código de producto. 16457657
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16457657 5 mg 5 miligramos
16424908 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16457657 Proveedor Medchem Express N.º de proveedor HY120012
MedChemExpress AZD9567 (compound 15) is a potent, oral active, non-steroidal and selective glucocorticoid receptor modulator (SGRM), with an IC50 of 3.8 nM. Exhibits excellent efficacy in the streptococcal cell wall (SCW) reactivation model of joint inflammation.
AZD9567 (compound 15) is a potent, oral active, non-steroidal and selective glucocorticoid receptor modulator (SGRM), with an IC50 of 3.8 nM. Exhibits excellent efficacy in the streptococcal cell wall (SCW) reactivation model of joint inflammation.
Especificaciones
Nombre del producto químico o material AZD9567
CAS 1893415-00-3
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C27H28F2N4O3
Cantidad 5 mg
Información de solubilidad DMSO : 100 mg/mL (202.21 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC(F)(F)C(N[C@H]([C@H](OC1=CC2=C(N(C(C=C3)=CN(C)C3=O)N=C2)C=C1)C4=CC=CC=C4)C(C)C)=O
Peso molecular (g/mol) 494.53
Formula Weight (peso de la fórmula) 494.53
Porcentaje de pureza 99.71%
Grado Research
Forma física Powder
Para utilizar con (aplicación) COVID-19-immunoregulation