Learn More
STEMCELL Technologies AZD8055
Descripción
- AZD8055 is a potent, selective ATP-competitive inhibitor of mammalian target of rapamycin (mTOR), with an IC50 value of 0.8 nM (Chresta et al.) It is very specific with ∼1,000-fold selectivity for mTOR over all PI3K isoforms and exhibits no activity against a panel of 260 kinases at concentrations up to 10 μM (Chresta et al.)
Especificaciones
Especificaciones
| CAS | 1009298-09-2 |
| Sinónimo | CCG-168 |
| Cantidad | 1 mg |
| Fórmula molecular | C25H31N5O4 |
| Pureza | ≥98% |