Learn More
Tocris Bioscience™ AZD 5248
Descripción
AZD 5248 is a potent cathepsin C (also known as dipeptidyl peptidase 1, or DPP1) inhibitor (IC50 values are 44 and 67 nM for human and rat, respectively). In vivo, AZD 5248 inhibition of cathepsin C correlates to the reduction of NSP (neutrophil serine protease) activity measured in rat bone marrow and blood. In rats, quantitative whole-body autoradiography studies demonstrate aortic binding of AZ 5248. This compound is orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-Amino-N-[(1S)-1-cyano-2-(4'-cyano[1,1'-biphenyl]-4-yl)ethyl]tetrahydro-2H-pyran-4-carboxamide |
| CAS | 1254318-44-9 |
| Cantidad | 1 mg |
| Objetivo | Synthetic Cathepsin Inhibitors |
| Fórmula molecular | C22H22N4O2 |
| Pureza | 0.98 |