Learn More
Tocris Bioscience™ AZD 1283
High affinity P2Y12 antagonist
Marca: Tocris Bioscience™ 6085/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
AZD 1283 is a potent and high affinity P2Y12 antagonist (IC50 = 11 nM in radioligand competition binding assay, and 25 nM in a GTPγS binding assay). Causes a dose-dependent increase in blood flow and inhibits ADP-induced platelet aggregation in vivo. Antithrombotic.Especificaciones
| Ethyl 5-cyano-2-methyl-6-[4-[[[(phenylmethyl)sulfonyl]amino]carbonyl]-1-piperidinyl]-3-pyridinecarboxylate | |
| Ethyl 5-cyano-2-methyl-6-[4-[[[(phenylmethyl)sulfonyl]amino]carbonyl]-1-piperidinyl]-3-pyridinecarboxylate | |
| Soluble to 100mM in DMSO | |
| White |
| 919351-41-0 | |
| 50 mg | |
| Solid | |
| Bioactive Small Molecule |
Título del producto
For Research Use Only.