Learn More
Tocris Bioscience™ AZ 5704
Descripción
AZ 5704 is a potent and selective ATM kinase inhibitor (IC50 = 0.6 nM in an enzyme inhibition assay). Exhibits > 600-fold selectivity for ATM over other kinases. Inhibits ATM kinase in an in vitro cellular assay (IC50 = 0.33 μM). Potentiates the antitumor effects of the topoisomerase 1 inhibitor irinotecan (CPT 11, Cat. No. 2688) in tumor bearing, immunocompromised mice. Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 7-Fluoro-6-[6-(methoxymethyl)-3-pyridinyl]-4-[[(1S)-1-(1-methyl-1H-pyrazol-3-yl)ethyl]amino]-3-quinolinecarboxamide |
| CAS | 1941214-06-7 |
| Cantidad | 25 mg |
| Fórmula molecular | C23H23FN6O2 |
| Formula Weight (peso de la fórmula) | 434.47 |
| Pureza | 98% |