Learn More
Tocris Bioscience™ AZ 1729
Descripción
AZ 1729 is a FFA2 allosteric agonist. Inhibits forskolin-induced cAMP increase and stimulates 35SGTPγS binding in biochemical assays (pEC50 values are 6.9 and 7.23, respectively). Binds at allosteric binding site. In functional assays, displays positive allosteric modulation of FFA-induced Gi signaling and negative allosteric modulation of FFA-induced Gq/G11 signaling. Inhibits lipolytic effect of isoproterenol (pEC50 = 5.03) and induces migration of human neutrophils in vitro.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-Fluoro-N-[3-[2-[(aminoiminomethyl)amino]-4-methyl-5-thiazolyl]phenyl]benzamide |
| CAS | 2016864-46-1 |
| Cantidad | 10 mg |
| Objetivo | Free Fatty Acid Receptor Agonists |
| Fórmula molecular | C18H16FN5OS |
| Formula Weight (peso de la fórmula) | 2323 |
| Pureza | 98% |