Learn More
Tocris™ AY 254
Descripción
AY 254 is a potent PAR2 biased agonist. Selectively activates ERK1/2 signaling (EC50 = 2 nM for ERK1/2 phosphorylation versus 80 nM for Ca2+ release). Reduces cytokine-induced caspase 3/8 activation, promotes scratch-wound healing, and induces IL-8 secretion, in human colorectal cancer (HT29) cells in vitro.
Especificaciones
Especificaciones
| CAS | 2093408-03-6 |
| Cantidad | 1 mg |
| Objetivo | Protease-Activated Receptor Agonists |
| Fórmula molecular | C30H49N9O6 |
| Pureza | 0.95 |