Learn More
Tocris Bioscience™ Avasimibe
Descripción
Avasimibe is an ACAT inhibitor (IC50 = 3.3 μM). Inhibits cholesterol esterification and reduces plasma cholesterol levels in rats. Also inhibits tumor growth and prolongs survival time in a melanoma mouse model and reduces tumor growth and metastasis in a pancreatic cancer mouse model.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2,6-Diisopropylphenyl (2-(2,4,6-triisopropylphenyl)acetyl)sulfamate |
| CAS | 166518-60-1 |
| Cantidad | 25 mg |
| Fórmula molecular | C29H43NO4S |
| Pureza | 98% |