Learn More
Atropine sulfate, MedChemExpress
Código de producto. 16408686
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
100 mg
Tamaño de la unidad:
100 miligramos
16408686 100 mg 100 miligramos
1 options
Código de producto. 16408686 Proveedor Medchem Express N.º de proveedor HYB1205A
MedChemExpress Atropine (Tropine tropate) sulfate is a competitive muscarinic acetylcholine receptor (mAChR) antagonist with IC50 values of 0.39 and 0.71 nM for Human mAChR M4 and Chicken mAChR M4, respectively. Atropine sulfate inhibits ACh-induced relaxations in human pulmonary veins. Atropine sulfate can be used for research of anti-myopia and bradycardia.
Atropine (Tropine tropate) sulfate is a competitive muscarinic acetylcholine receptor (mAChR) antagonist with IC50 values of 0.39 and 0.71 nM for Human mAChR M4 and Chicken mAChR M4, respectively. Atropine sulfate inhibits ACh-induced relaxations in human pulmonary veins. Atropine sulfate can be used for research of anti-myopia and bradycardia.
Especificaciones
Nombre del producto químico o material Atropine sulfate
CAS 55-48-1
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C17H23NO3.1/2H2SO4
Cantidad 100 mg
Sinónimo Tropine tropate sulfateDL-Hyoscyamine sulfate Sulfatropinol
Información de solubilidad H2O : 100 mg/mL (295.50 mM; Need ultrasonic) ∣DMSO : 62.5 mg/mL (184.69 mM; Need ultrasonic)
SMILES O=C(O[C@@H]1C[C@@H](N2C)CC[C@@H]2C1)C(CO)C3=CC=CC=C3.[0.5H2SO4]
Peso molecular (g/mol) 338.41
Formula Weight (peso de la fórmula) 338.41
Porcentaje de pureza 98.07%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Neuroscience-Neuromodulation