Learn More
ATP5D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16883784 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16883784 20 μL 20 microlitros
16873784 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16883784 Proveedor Proteintech N.º de proveedor 148931AP20UL
Rabbit Polyclonal Antibody
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the delta subunit of the catalytic core. Alternatively spliced transcript variants encoding the same isoform have been identified.
Especificaciones
Antígeno ATP5D
Aplicaciones Western Blot, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.14 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica ATP5D
N.º de referencia del gen P30049, P35434, Q9D3D9
Alias de gen ATP5D, F ATPase delta subunit
Símbolos de los genes ATP5D
Especie del huésped Rabbit
Inmunógeno ATP5D Fusion Protein Ag6682
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 245965, 513, 66043
Especies diana Mouse, Rat, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG