Learn More
ATF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16877483 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16877483 20 μL 20 microlitros
16867483 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16877483 Proveedor Proteintech N.º de proveedor 108351AP20UL
Rabbit Polyclonal Antibody
This antibody is a rabbit polyclonal antibody raised against full length human ATF4 antigen. The antibody recognizes the 38 kDa ATF4 protein and its phosphorylated forms (50 kDa). ATF4 can bind DNA as a homodimer and as a heterodimer. ATF4 is ubiquitinate
This gene encodes a transcription factor that was originally identified as a widely expressed mammalian DNA binding protein that could bind a tax-responsive enhancer element in the LTR of HTLV-1. The encoded protein was also isolated and characterized as the cAMP-response element binding protein 2 (CREB-2). The protein encoded by this gene belongs to a family of DNA-binding proteins that includes the AP-1 family of transcription factors, cAMP-response element binding proteins (CREBs) and CREB-like proteins. These transcription factors share a leucine zipper region that is involved in protein-protein interactions, located C-terminal to a stretch of basic amino acids that functions as a DNA binding domain. Two alternative transcripts encoding the same protein have been described. Two pseudogenes are located on the X chromsome at q28 in a region containing a large inverted duplication.
Especificaciones
Antígeno ATF4
Aplicaciones Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation
Clasificación Polyclonal
Concentración 0.27 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica ATF4
N.º de referencia del gen P18848, Q06507, Q9ES19
Alias de gen ATF4, CREB 2, CREB2, DNA binding protein TAXREB67, TAXREB67, TXREB
Símbolos de los genes ATF4
Especie del huésped Rabbit
Inmunógeno ATF4 Fusion Protein Ag1279
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11911, 468, 79255
Especies diana Human, Rat, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG