Learn More
Astemizole, 99+%, Thermo Scientific™
Marca:  Thermo Scientific Alfa Aesar J60339.MF
Cantidad 50mg
Código de producto. 15423119
  • 164.00€ / 50 miligramos
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
It is soluble in DMSO (25 mg/ml ), ethanol (25 mg/ml), chloroform, methanol, and water (partly miscible).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storag
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Astemizole
C28H31FN4O
50mg
astemizole, hismanal, histaminos, paralergin, laridal, retolen, alermizol, astemison, astemizol, histamen
GXDALQBWZGODGZ-UHFFFAOYSA-N
1-[(4-fluorophenyl)methyl]-N-[1-[2-(4-methoxyphenyl)ethyl]piperidin-4-yl]benzimidazol-2-amine
2247
458.57
68844-77-9
MFCD00153919
14,856
It is soluble in DMSO (25mg/ml ),ethanol (25mg/ml),chloroform,methanol,and water (partly miscible).
COC1=CC=C(C=C1)CCN2CCC(CC2)NC3=NC4=CC=CC=C4N3CC5=CC=C(C=C5)F
458.581
CHEBI:2896
≥99%
Sugerencias de productos
Sugerencias de productos
Videos
SDS Documentos