Learn More
Tocris Bioscience™ ASP 7967
Descripción
ASP 7967 is a ligand for RNA aptamer AC17-4 that activates gene expression in synthetic aptazime-based riboswitches at as low as 5 μM concentration, without relying on exogenous protein factors. ASP 7967 binds aptamer AC17-4 with high affinity in HEK293 cells (Kd = 12 nM; EC50 = 2.3 μM), activating gene expression (>10-fold) in HEK293 cells. ON/OFF ratio of riboswitches activated by ASP 7967 approaches 300, with a low basal expression level in cultured cells.
In vivo, ASP 7967 upregulates the serum hEPO concentration by 7.2-fold over the vehicle control (mice). Cell permeable and low-toxic, ASP 7967 is orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N2,N4-Bis(4-fluorophenyl)-N6-(2-pyrimidinylmethyl)-1,3,5-triazine-2,4,6-triamine hemifumarate |
| CAS | 1246958-42-8 |
| Cantidad | 10 mg |
| Objetivo | DNA, RNA and Protein Synthesis Compounds |
| Fórmula molecular | C20H16F2N8.½C4H4O4 |
| Pureza | 98% |