Learn More
Tocris Bioscience™ AS 1842856
Descripción
AS 1842856 is a potent and selective Foxo1 inhibitor (IC50 values are 33 nM at Foxo1 and >1 μM at Foxo3a and Foxo4); decreases Foxo-mediated promoter activity by 70%, 20% and 3% at Foxo1, Foxo4 and Foxo3, respectively. Regulates gluconeogenesis both in vitro and in vivo. Attenuates fasting plasma glucose level in diabetic db/db mice. Also suppresses autophagy, adipocyte differentiation and soluble guanylyl cyclase expression. Also inhibits STAT3 signaling in NSCLC. Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 5-Amino-7-(cyclohexylamino)-1-ethyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid |
| CAS | 836620-48-5 |
| Cantidad | 10 mg |
| Fórmula molecular | C18H22FN3O3 |
| Pureza | 98% |