missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARHGAP18 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15704701 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Unit Size:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
15704701 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15704701 Supplier Invitrogen™ Supplier No. PA556790

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: STTIKVMEKP PFDRSISQDS LDELSMEDYW IELENIKKSS ENSQEDQEVV VVKEPDEGEL EEEWLKEAGL SNLFGESAGD PQESI Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.

ARHGAP18 is one member of the human RhoGAP family with approximately 80 RhoGAP proteins known to be encoded in the human genome. Rho proteins belong to the Ras superfamily that is composed of over 50 members divided into six families, including Ras, Sar, Rho, Ran, Rab and Arf. Rho GTPases are important regulators of the actin cytoskeleton and consequently influence the shape and migration of cells. ARHGAP18 is linked to Ras, and thus, to EGFR-mediated proliferation, migration and differentiation. ARHGAP18 is precisely contained within chromosome 6q22-24, which has been shown to be linked to schizophrenia, suggesting that ARHGEP18 may play a role in this condition.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ARHGAP18
Aplicaciones Immunohistochemistry (Paraffin), Immunocytochemistry
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica ARHGAP18
N.º de referencia del gen Q8N392
Alias de gen 4833419J07Rik; AI314906; AI843501; Arhgap18; bA307O14.2; MacGAP; Rho GTPase activating protein 18; rho GTPase-activating protein 18; Rho-type GTPase-activating protein 18; SENEX
Símbolos de los genes ARHGAP18
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human ARHGAP18. Recombinant protein control fragment (Product #RP-94893).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 93663
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.