Learn More
Arginase 1/ARG1/liver Arginase Antibody, Novus Biologicals™
Descripción
Arginase 1/ARG1/liver Arginase Polyclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Peptide ELISA.
Especificaciones
Especificaciones
| N.º de referencia del gen | P07824 |
| Símbolos de los genes | ARG1 |
| Peso molecular del antígeno | 37 kDa |