Learn More
Tocris Bioscience™ AR-C 141990 hydrochloride
MCT1 inhibitor
Marca: Tocris Bioscience™ 5658/1
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
AR-C 141990 hydrochloride is a monocarboxylate transporter (MCT1) inhibitor (pKi = 7.6. Exhibits approximately 10-fold selectivity for MCT1 over MCT2 and no significant activity against MCT3 or 4. Inhibits graft versus host response and prolongs cardiac graft survival in a rat model. Immunosuppressant. Also inhibits brain penetration of GHB analog HOCPCA.Especificaciones
| 5-[[(3R)-3-Hydroxy-1-pyrrolidinyl]carbonyl]-3-methyl-1-(2-methylpropyl)-6-(4-quinolinylmethyl)thieno[2,3-d]pyrimidine-2,4(1H,3H)-dione | |
| 2250019-94-2 | |
| >98% | |
| 1 mg |