Learn More
Tocris Bioscience™ AR 420626
Descripción
AR 420626 is a FFA3 agonist (pEC50 = 5.74). Inhibits nicotine and serotonin induced changes in motility of isolated muscle strips from rat colon. Also suppresses serotonin-induced fecal output in rats.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-(2,5-Dichlorophenyl)-4-(furan-2-yl)-2-methyl-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carboxamide |
| CAS | 1798310-55-0 |
| Intervalo de porcentaje de ensayo | >98% |
| Cantidad | 25 mg |
| Fórmula molecular | C21H18Cl2N2O3 |
| Formula Weight (peso de la fórmula) | Observed MW: 417.29 |