Learn More
Tocris Bioscience™ AP 1903
Descripción
AP 1903 is a chemical protein dimerizer for use in FKBP fusion protein systems. Induces apoptosis in cells expressing a fusion protein consisting of FKBPF36V and intracellular domain of Fas receptor with high potency (EC50 = 0.1 nM). Exhibits >50-fold selectivity for FKBPF36V-Fas fusion protein over wild-type FKBP. Also induces apoptosis in cells expressing Cas9-FK506 fusion proteins, in inducible caspase 9 (iCas9) suicide gene system. Active in vivo.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2,2 '-[1,2-Ethanediylbis[imino(2-oxo-2,1-ethanediyl)oxy-3,1-phenylene[(1 R)-3-(3,4-dimethoxyphenyl)propylidene]]] bis[(2 S)-1-[(2 S)-1-oxo-2-(3,4,5-trimethoxyphenyl)butyl]-2-piperidinecarboxylate] |
| CAS | 195514-63-7 |
| Cantidad | 5 mg |
| Objetivo | Protein Dimerizers |
| Fórmula molecular | C78H98N4O20 |
| Pureza | 0.98 |