Learn More
vav 1 guanine nucleotide exchange factor, Mouse, Clone: 1A6, Abnova™
Descripción
The protein encoded by this proto-oncogene is a member of the Dbl family of guanine nucleotide exchange factors (GEF) for the Rho family of GTP binding proteins. The protein is important in hematopoiesis, playing a role in T-cell and B-cell development and activation. This particular GEF has been identified as the specific binding partner of Nef proteins from HIV-1. Coexpression and binding of these partners initiates profound morphological changes, cytoskeletal rearrangements and the JNK/SAPK signaling cascade, leading to increased levels of viral transcription and replication. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | vav 1 guanine nucleotide exchange factor |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 1A6 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against a partial recombinant VAV1. |
| Formulación | PBS with no preservative; pH 7.4 |
| génica | VAV1 |
| N.º de referencia del gen | BC013361 |
| Alias de gen | VAV |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.