Learn More
TRA-1-60 Antigen Mouse anti-Human, Alexa Fluor 647, Clone: TRA-1-60, BD
Descripción
The TRA-1-60 monoclonal antibody reacts with the neuraminidase-resistant form of a pluripotent-stem-cell-specific epitope on a high-molecular-weight transmembrane glycoprotein. The TRA-1-60 antigen is a sialylated epitope on the same keratan sulfate core molecule, podocalyxin, as 4 other distinct antigens on tumor-derived cell lines, TRA-1-81, GCTM2, K4, and K21. The expression of TRA-1-60 antigen is stage-specific and can be used to characterize embryonic cells and monitor their differentiation. The antigen is found on teratocarcinoma (embryonal carcinoma or EC), embryonic inner cell mass (but not morula or trophoblast), and embryonic stem (ES) cells. TRA-1-60 antigen is released into the serum of patients bearingtesticular tumors containing EC cells. As human EC and ES cells undergo differentiation, expression of TRA-1-60 antigen is lost. Expression of TRA-1-60 antigen has also been observed on a rhesus monkey ES cell line (Thomson et al, 1995).
Especificaciones
Especificaciones
| Antígeno | TRA-1-60 Antigen |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | TRA-1-60 |
| Conjugado | Alexa Fluor 647 |
| Descripción | TRA-1-60(R) |
| Formulación | Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human Embryonal Carcinoma Cell Line |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.