missing translation for 'onlineSavingsMsg'
Learn More

anti-TLR3/CD283 PE, Clone: TLR3.7, Invitrogen™

Código de producto. 15394196
Cambiar vista
Click to view available options
Cantidad:
25 μg
Tamaño de la unidad:
25 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15394196 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15394196 Proveedor Invitrogen N.º de proveedor A18430

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

TLR3/CD283 Monoclonal antibody A18430 from Invitrogen specifically detects TLR3/CD283 in Human samples. It is validated for Flow Cytometry.

TLR3 is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression.
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG1, κ
Clone: TLR3.7
Immunogen: human CD283
Formulation: 25μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: Toll-like Receptor 3, IIAE2
Uniprot Accession: O15455
Entrez GeneID: 7098
Gene Symbol: TLR3
This product detects TLR3/CD283 in Human samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is human CD283.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TLR3/CD283
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon TLR3.7
Conjugado PE
Formulación 25μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
N.º de referencia del gen O15455
Alias de gen Toll-like Receptor 3, IIAE2
Símbolos de los genes TLR3
Especie del huésped Mouse
Inmunógeno human CD283
Cantidad 25 μg
Primario o secundario Primary
ID de gen (Entrez) 7098
Especies diana Human
Contenido y almacenamiento Store at 4° C
Formulario Purified
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.