Learn More
PSD-95 Mouse, Unlabeled, Clone: 16, BD
Código de producto. 15432233
Change view
Click to view available options
Cantidad:
150 μg
50 μg
Tamaño de la unidad:
1 set
cada uno
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
15432233 50 μg cada uno
15855489 150 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 15432233 Proveedor BD Biosciences N.º de proveedor 610495
Mouse Monoclonal Antibody
PostSynaptic Density (PSD) refers to a dense region of submembranous cytoskeleton found most prominently in postsynaptic membranes of the CNS. Possible functions for the PSD include regulation and aggregation of receptors,structural stabilization of the synaptic junction, and transduction of signals from membrane receptors. Some of the proteins associated with the PSD are fodrin, tubulin, calmodulin, CaM Kinase II, PSD-95, and PSD-93. PSD-95 (SAP90) is a protein that interacts with the NMDA receptor NMDAR2B, neuronal NOS(nNOS or bNOS), and other proteins. PSD-95 contains one SH3 domain in its carboxy-terminal domain, as well as three conserved repeat regions called GLGF or PDZ domains. nNOS, which is concentrated in synaptic junctions, also contains a PZD domain. PSD-95 and nNOS interact via their respective PZD domains, which may mediate the binding of nNOS to skeletal muscle syntrophin.
This antibody is routinely tested by western blot analysis. Other applications were tested in BD Biosciences Pharmingen during antibody development only or reported in the literature.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno PSD-95
Aplicaciones Western Blot
Clasificación Monoclonal
Clon 16
Concentración 250μg/mL
Conjugado Unconjugated
Descripción PostSynaptic Density-95; SAP90
Formulación Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Método de purificación Affinity Purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Mouse, Rat
Contenido y almacenamiento Store undiluted at -20°C.
Isotype IgG1
Mostrar más Mostrar menos
For Research Use Only.