Learn More
Peptidoglycan Monoclonal Antibody (3F6B3 (10H6)), Invitrogen™
Mouse Monoclonal Antibody
Marca:  Invitrogen MA516509
Detalles adicionales : Peso : 0.01000kg
Código de producto. 15159335
  • 588.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock
Descripción
Descripción
Treatment with strong acid (Gram positive bacteria) or with a detergent such as SDS (Gram-negative bacteria) may be necessary to expose the epitope.
Peptidoglycan Monoclonal antibody specifically detects Peptidoglycan in Bacteria samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).
Especificaciones
Especificaciones
Peptidoglycan
Monoclonal
Conc. Not Determined
ascites with no preservative
Mouse
100 μL
Primary
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
3F6B3 (10H6)
Unconjugated
PG
This antibody was raised against insoluble peptidoglycan obtained by TCA-heat and ethanol extraction of Streptococcus mutans BHT cells.
RUO
Bacteria
Antibody
IgG1