Learn More
PAC-1 Mouse anti-Human, FITC, Clone: PAC-1, BD
Código de producto. 15865898
Change view
Click to view available options
Cantidad:
50 Tests
Tamaño de la unidad:
cada uno
15865898 50 Tests cada uno
1 options
Código de producto. 15865898 Proveedor BD Biosciences N.º de proveedor 340507
Mouse Monoclonal Antibody
PAC-1 recognizes an epitope on the glycoprotein IIb/IIIa (gpIIb/IIIa, aIIbb3) complex of activated platelets at or near the platelet fibrinogen receptor. The gpIIb/IIIa complex is a member of a family of heterodimeric adhesive protein receptors found on a variety of cell types that have been named the integrin or the cytoadhesion family.The gpIIb/IIIa complex is located on the surface membrane of resting platelets. Platelet activation induces a calcium-dependent conformational change in gpIIb/IIIa that exposes a ligand binding site. Four adhesive macromolecules are capable of interacting with the activated form of gpIIb/IIIa: fibrinogen, von Willebrand factor (vWf), fibronectin, and vitronectin. The binding of fibrinogen to the gpIIb/IIIa receptor is required for platelet aggregation. PAC-1 binds only to activated platelets and appears to be specific for this recognition site within gpIIb/IIIa. Approximately 45,000 to 50,000 gpIIb/IIIa receptors appear on the platelet surface upon activation. PAC-1 inhibits fibrinogen- mediated platelet aggregation.
Flow Cytometry
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno PAC-1
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon PAC-1
Concentración 25μg/mL
Conjugado FITC
Formulación Phosphate buffered saline with gelatin and 0.1% sodium azide.
Especie del huésped Mouse
Inmunógeno Human platelets
Cantidad 50 Tests
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgM κ
Mostrar más Mostrar menos
For Research Use Only.