Learn More
nonmetastatic cells 1, protein (NM23A) expressed in, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Descripción
This gene (NME1) was identified because of its reduced mRNA transcript levels in highly metastatic cells. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by this gene) and 'B' (encoded by NME2) isoforms. Mutations in this gene have been identified in aggressive neuroblastomas. Two transcript variants encoding different isoforms have been found for this gene. Co-transcription of this gene and the neighboring downstream gene (NME2) generates naturally-occurring transcripts (NME1-NME2), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product. [provided by RefSeq]
Especificaciones
Especificaciones
| Antígeno | non-metastatic cells 1, protein (NM23A) expressed in |
| Aplicaciones | Immunofluorescence, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against a full-length human NME1 protein. |
| Formulación | PBS with no preservative; pH 7.4 |
| génica | NME1 |
| N.º de referencia del gen | NM_198175 |
| Alias de gen | AWD/GAAD/NB/NBS/NDPK-A/NDPKA/NM23/NM23-H1 |
| Símbolos de los genes | NME1 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.