missing translation for 'onlineSavingsMsg'
Learn More

MAPKAPK-2 Mouse, Alexa Fluor 488, Clone: P24-694, BD

Código de producto. 15871949
Change view
Click to view available options
Cantidad:
50 Tests
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15871949 50 Tests cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15871949 Proveedor BD Biosciences N.º de proveedor 562471

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The P24-694 monoclonal antibody specifically binds to the phosphorylated T334 site (pT334) of MAPKAPK-2. MAPKAPK-2 is a serine/threonine protein kinase. This ∽49 kDa member of the MAPKAPK family of protein kinases is also known as mitogen-activated protein kinase-activated protein kinase 2. MAPKAPK-2 is phosphorylated and activated by p38 MAP kinase in response to stress, cytokines and chemokines. MAPKAPK-2 is phosphorylated on multiple sites including Thr222, Ser272 and Thr334. Phosphorylation of any two of these three amino acid residues seems to be required for the activation of this kinase thatserves multiple cellular functions. Phosphorylation of Thr334 was reported to be essential for nuclear export of the heterodimer formed between p38 MAPK and MAPKAPK-2. Mice deficient in MAPKAPK-2 have been shown to be protected from ischemic injury. MAPKAPK-2 is also reported to serve as a cell cycle checkpoint kinase in response to UV irradiation. The heat shock protein, HSP27 was shown to be one of the major substrates of MAPK and MAPKAPK-2.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MAPKAPK-2
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon P24-694
Conjugado Alexa Fluor 488
Descripción MAP kinase-activated protein kinase 2, MAPKAP kinase 2; MK2; MAPK2
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Phosphorylated human MAPKAPK-2 peptide
Método de purificación Affinity Purified
Cantidad 50 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human, Mouse
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.