missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LINGO-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-46335
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
LINGO-1 Polyclonal specifically detects LINGO-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Especificaciones
| LINGO-1 | |
| Polyclonal | |
| Unconjugated | |
| Whole antisera with No Preservative | |
| LINGO1 | |
| A synthetic peptide fromc-terminal region of human Lingo1 conjugated to an immunogenic carrier protein was used as the antigen. The peptide is homologous in many other species. | |
| 0.1 mL | |
| Primary | |
| Specific for Lingo1. | |
| Human, Mouse, Rat | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| LYOPH | |
| Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| FLJ14594, LERN1UNQ201, leucine rich repeat and Ig domain containing 1, leucine rich repeat neuronal 6A, Leucine-rich repeat and immunoglobilin domain-containing protein 1, Leucine-rich repeat neuronal protein 1, Leucine-rich repeat neuronal protein 6A, LRRN6Aleucine-rich repeat neuronal 6A, MGC17422 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| 84894 | |
| Reconstitute with 0.1 ml sterilized water to desired concentration. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
For Research Use Only