missing translation for 'onlineSavingsMsg'
Learn More

Ikaros Mouse anti-Human, Alexa Fluor 647, Clone: R32-1149, BD

Código de producto. 17277474
Change view
Click to view available options
Cantidad:
50 Tests
Tamaño de la unidad:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17277474 50 Tests 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17277474 Proveedor BD Biosciences N.º de proveedor 564868

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The R32-1149 monoclonal antibody specifically binds to Ikaros, which is encoded by IKZF1 (IKAROS family zinc finger 1). Ikaros is also known as Lymphoid transcription factor LyF-1 (LYF1) and ZNFN1A1 [Zinc finger protein, subfamily 1A, 1 (Ikaros)]. Ikaros belongs to the IKAROS family of zinc-finger transcription factors. This DNA-binding transcription factor forms homodimers and heterodimers with other IKAROS family members. It is involved in the development of lymphoid tissues and remains abundantly expressed in the thymus and by cells within peripheral lymphoid tissues. Ikaros functions in the maturation, differentiation, and homeostasis of various hematopoietic cells including T lymphocytes, B cells, NK cells, and neutrophils. Ikaros regulates the expression of a number of genes including various genes expressed during early stages of B- and T-cell development. Several alternatively-spliced human Ikaros isoforms have been described. Abnormal expression of different Ikaros isoforms has been associated with hematologic malignancies . Flow cytometric analysis of 293F cells transfected with members of the IKAROS family has demonstrated that clone R32-1149 is negative on Helios, Aiolos, and Eos transfectants.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Ikaros
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon R32-1149
Conjugado Alexa Fluor 647
Descripción IKZF1; LYF1; hIk-1; IK1; ZNFN1A1
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Especie del huésped Mouse
Inmunógeno Human Ikaros Recombinant Protein
Método de purificación Affinity Purified
Cantidad 50 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.