Learn More
FR4 (Folate receptor 4) Rat anti-Mouse, APC, Clone: 12A5, BD
Descripción
The monoclonal antibodies TH6 and 12A5 recognize Folate Receptor 4 (FR4), also known as the membrane folate-binding protein 3 (FBP3). FR4 is a heavily glycosylated 35kDa receptor expressed exclusively in lymphoid tissue and an isoform of the family of receptors that recognize the essential nutrient folic acid. Natural T regulatory cells constitutively express high levels of FR4. Differential expression of FR4 in combination with CD25 can distinguish four functionally distinct CD4+ T cell subpopulations; Natural Tregs, effector T cells, memory-like T cells and Nanve T cells. FR4hi CD25+ expressing CD4+ T cells also express high amounts of Foxp3, GITR and CTLA-4. Monoclonal antibody TH6 and 12A5 stained CD25+CD4+ T cells at a higher level than other CD4+ or CD8+ T cells. In addition, in vivo injection of TH6 monoclonal antibody reduced the number of CD25+CD4+ T cells and CD25-CD4+ T cells in peripheral blood. Clone 12A5 has been demonstrated to work in western blot. Clones TH6 and 12A5 do not block one another in a flow cytometric assay.
Host Species: Rat
Clone: 12A5
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Mouse CD4+ CD25+ T cells
Flow Cytometry
Especificaciones
Especificaciones
| Antígeno | FR4 (Folate receptor 4) |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 12A5 |
| Concentración | 0.2mg/mL |
| Conjugado | APC |
| Descripción | Folate receptor 4, FBP, FBP3, FRdelta, FRd |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Especie del huésped | Rat |
| Inmunógeno | Mouse CD4+ CD25+ T cells |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.