missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Fc epsilon Receptor I alpha/FceR1 PE, Clone: AER-37, Invitrogen™
Descripción
Fc epsilon Receptor I alpha/FceR1 Monoclonal antibody A18416 from Invitrogen specifically detects Fc epsilon Receptor I alpha/FceR1 in Human samples. It is validated for Flow Cytometry.
Fc epsilonR1 alpha is a subunit of the high affinity receptor of IgE to which it directly binds. Fc epsilonR1 alpha is a tetrameric complex consisting of one alpha, one ß and two additional subunits. The latter two are required for signal transduction activity. The Fc epsilonR1 complex plays an important role in triggering allergic responses.
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG2b, κ
Clone: AER-37
Immunogen: alpha subunit of human Fc epsilon receptor I (FceR1)
Formulation: 25 Tests Purified Antibody (0.05mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: High affinity immunoglobulin epsilon receptor subunit alpha, Fc-epsilon RI-alpha, FcERI, IgE Fc receptor subunit alpha, FCER1A, FCE1A
Uniprot Accession: P12319
Entrez GeneID: 2205
Gene Symbol: FCER1A
This product detects Fc epsilon Receptor I alpha/FceR1 in Human samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is alpha subunit of human Fc epsilon receptor I (FceR1).
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG2b, κ
Clone: AER-37
Immunogen: alpha subunit of human Fc epsilon receptor I (FceR1)
Formulation: 25 Tests Purified Antibody (0.05mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: High affinity immunoglobulin epsilon receptor subunit alpha, Fc-epsilon RI-alpha, FcERI, IgE Fc receptor subunit alpha, FCER1A, FCE1A
Uniprot Accession: P12319
Entrez GeneID: 2205
Gene Symbol: FCER1A
This product detects Fc epsilon Receptor I alpha/FceR1 in Human samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is alpha subunit of human Fc epsilon receptor I (FceR1).
Especificaciones
Especificaciones
| Antígeno | Fc epsilon Receptor I alpha/FceR1 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | AER-37 |
| Conjugado | PE |
| Formulación | 25 Tests Purified Antibody (0.05mg/ml) in proprietary buffer, with 0.09% sodium azide |
| N.º de referencia del gen | P12319 |
| Alias de gen | High affinity immunoglobulin epsilon receptor subunit alpha, Fc-epsilon RI-alpha, FcERI, IgE Fc receptor subunit alpha, FCER1A, FCE1A |
| Símbolos de los genes | FCER1A |
| Especie del huésped | Mouse |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?